Learn More
Inorganic Ventures SPECTRO GENESIS ICAL STD 125mL
SDP
Catalog No. 50723546
Encompass_Preferred
Matrix: Nitric Acid / Hydrochloric Acid / Hydrofluoric Acid (HNO3 / HCl / HF), Instrument(s): Spectro, Analyte: Be, Sr, and Li 2 ug/mL each, Ce, Eu, Fe, Ni, K, Si, Ti, Y, Cu, In, P, V, and Zr 10 ug/mL each, Mn, Mo, Sc, and Na 5 ug/mL each, Ca 1 ug/mL, and S 50 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-546 Supplier Inorganic Ventures Supplier No. GENESISICAL125ML
Matrix: Nitric Acid / Hydrochloric Acid / Hydrofluoric Acid (HNO3 / HCl / HF), Instrument(s): Spectro, Analyte: Be, Sr, and Li 2 ug/mL each, Ce, Eu, Fe, Ni, K, Si, Ti, Y, Cu, In, P, V, and Zr 10 ug/mL each, Mn, Mo, Sc, and Na 5 ug/mL each, Ca 1 ug/mL, and S 50 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search