Learn More
Medchemexpress LLC ICAM-1-IN-1 50mg | 50mg
SDP
Catalog No. 5000321895
Encompass_Preferred
ICAM-1-IN-1 is a potent and selective inhibitor of E-selectin and ICAM-1 with IC50 values of 7 and 5 nM respectively
Catalog No. 50-003-21895 Supplier Medchemexpress LLC Supplier No. HYU0000350MG
ICAM-1-IN-1 is a potent and selective inhibitor of E-selectin and ICAM-1 with IC50 values of 7 and 5 nM respectively