Learn More
Aviva Systems Biology Corp ICAM1 Antibody - N-terminal region (ARP59129_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59129_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ICAM1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ICAM1. Alternate names: BB2, CD54, P3.58, ICAM-1, ICAM1ApplicationWB. Species reactivity: Human, Rabbit
Catalog No. 50-286-589