Learn More
ACROBiosystems Recombinant Protein;Human ICAM-1 / CD54 Protein, His Tag;HEK293;1MG;IC1,ICAM1,ICAM-1,BB2,BB-2,CD54,CD-54,P3.58
Catalog No. 501658324
Encompass_Preferred
Recombinant Protein;Human ICAM-1 / CD54 Protein, His Tag;1MG;Human ICAM-1, His Tag (IC1-H52H5) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Glu 480 (Accession # P05362-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 51.4 kDa. The protein migrates as 60-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8324 Supplier ACROBiosystems Supplier No. IC1H52H51MG
Recombinant Protein;Human ICAM-1 / CD54 Protein, His Tag;1MG;Human ICAM-1, His Tag (IC1-H52H5) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Glu 480 (Accession # P05362-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 51.4 kDa. The protein migrates as 60-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.