Learn More
Glass Expansion RF Coil, Glass Expansion, silver plated and Teflon-coated, for iCAP 6000/7000 Mark II. For enhanced RF conductivity. Installion tool required (not included)
SDP
Catalog No. 501694676
Encompass_Preferred
RF Coil Silver and PTFE Sheathed for Thermo 6000/7000 Mark II ICP-OES. Compatible with Thermo 6000/7000 Mark II ICP-OES. The RF coil is susceptible to corrosion in the torch box, which eventually lowers the conductivity of the coil. The greater the corrosion, the larger amount of energy is needed to produce a plasma of the same power. Coils in a corroded condition, increases the stress applied to other components in the RF generating system, and can shorten the lifetime of your torch. Copper is used as the base material of most commercially available coils but different plating materials are available. Silver has the best conductance & gold the best corrosion resistance. Only Silver is available for this ICP.To install your coil and ensure proper alignment, we highly recommend our re-useable RF Coil Installation Tool Kit, 70-803-0960, which is now obsolete.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-169-4676 Supplier Glass Expansion Supplier No. 709004005S
RF Coil Silver and PTFE Sheathed for Thermo 6000/7000 Mark II ICP-OES. Compatible with Thermo 6000/7000 Mark II ICP-OES. The RF coil is susceptible to corrosion in the torch box, which eventually lowers the conductivity of the coil. The greater the corrosion, the larger amount of energy is needed to produce a plasma of the same power. Coils in a corroded condition, increases the stress applied to other components in the RF generating system, and can shorten the lifetime of your torch. Copper is used as the base material of most commercially available coils but different plating materials are available. Silver has the best conductance & gold the best corrosion resistance. Only Silver is available for this ICP.To install your coil and ensure proper alignment, we highly recommend our re-useable RF Coil Installation Tool Kit, 70-803-0960, which is now obsolete.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001