Learn More
Glass Expansion RF Coil, Glass Expansion, silver plated and Teflon-coated, for iCAP 6000 Mark I. For enhanced RF conductivity. Installion tool required (not included)
SDP
Catalog No. 501694674
Encompass_Preferred
RF Coil Silver for Thermo 6000 Mark I ICP-OES. Compatible with Thermo 6000 Mark I ICP-OES. The RF coil is susceptible to corrosion in the torch box, which eventually lowers the conductivity of the coil. The greater the corrosion, the larger amount of energy is needed to produce a plasma of the same power. Coils in a corroded condition, increases the stress applied to other components in the RF generating system, and can shorten the lifetime of your torch. Copper is used as the base material of most commercially available coils but different plating materials are available. Silver has the best conductance & gold the best corrosion resistance. Only Silver is available for this ICP.To install your coil and ensure proper alignment, we highly recommend our re-useable RF Coil Installation Tool Kit (NC2081827).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-169-4674 Supplier Glass Expansion Supplier No. 709004002S
RF Coil Silver for Thermo 6000 Mark I ICP-OES. Compatible with Thermo 6000 Mark I ICP-OES. The RF coil is susceptible to corrosion in the torch box, which eventually lowers the conductivity of the coil. The greater the corrosion, the larger amount of energy is needed to produce a plasma of the same power. Coils in a corroded condition, increases the stress applied to other components in the RF generating system, and can shorten the lifetime of your torch. Copper is used as the base material of most commercially available coils but different plating materials are available. Silver has the best conductance & gold the best corrosion resistance. Only Silver is available for this ICP.To install your coil and ensure proper alignment, we highly recommend our re-useable RF Coil Installation Tool Kit (NC2081827).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001