Learn More
Thermo Scientific Icap 6000 Multi Element Test 1 Ea
Catalog No. NC1662133 Shop All Thermo Scientific Products
Encompass
Multi-element test accessory for optical emission spectrometry using inductively coupled plasma allows running multi-element calibrations and samples for elemental analysis™
Catalog No. NC1662133 Supplier Thermo Scientific Supplier No. 430122821401
Multi-element test accessory for optical emission spectrometry using inductively coupled plasma allows running multi-element calibrations and samples for elemental analysis™