Learn More
Glass Expansion Quartz Torch for iCAP 6000/7000 Duo (842312051841)
SDP
Catalog No. NC1417608
Encompass_Preferred
Quartz EMT Torch, standard for Thermo Fisher Scientific DUO 6000 and 7000 Series ICP-OES (842312051841).For samples containing HF or volatile organics see catalog number 50-169-4650.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. NC1417608 Supplier Glass Expansion Supplier No. 318082811
Quartz EMT Torch, standard for Thermo Fisher Scientific DUO 6000 and 7000 Series ICP-OES (842312051841).For samples containing HF or volatile organics see catalog number 50-169-4650.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001