Learn More
Medchemexpress LLC Icapamespib hydrochloride | 2267287-26-1 | 98.7% | C19H25Cl2IN6O2S | 10MG
SDP
Catalog No. 5000259632
Encompass_Preferred
Icapamespib hydrochloride is the hydrochloride salt of icapamespib, a selective, orally active inhibitor of epichaperomes assembled by HSP90. It can cross the blood-brain barrier and promotes epichaperome disassembly by non-covalently binding HSP90, supporting studies of protein-protein interaction networks in neurodegenerative disease and oncology research.
  • Selective inhibitor of epichaperomes assembled by HSP90.
  • Orally active compound with slow dissociation kinetics.
  • Crosses the blood-brain barrier for central nervous system research.
  • Applicable to neurodegenerative disease and cancer research models.
  • Supplied as a white to off-white solid with high purity for research use.
Catalog No. 50-002-59632 Supplier Medchemexpress LLC Supplier No. HY137441A10MG
Icapamespib hydrochloride is the hydrochloride salt of icapamespib, a selective, orally active inhibitor of epichaperomes assembled by HSP90. It can cross the blood-brain barrier and promotes epichaperome disassembly by non-covalently binding HSP90, supporting studies of protein-protein interaction networks in neurodegenerative disease and oncology research.
  • Selective inhibitor of epichaperomes assembled by HSP90.
  • Orally active compound with slow dissociation kinetics.
  • Crosses the blood-brain barrier for central nervous system research.
  • Applicable to neurodegenerative disease and cancer research models.
  • Supplied as a white to off-white solid with high purity for research use.