Learn More
Medchemexpress LLC Icapamespib hydrochloride | 2267287-26-1 | 98.74% | 599.32 | 25 MG
SDP
Catalog No. 5000350673
Encompass_Preferred
Icapamespib hydrochloride is a selective, orally active inhibitor of Epichaperomes assembled by HSP90 with slow dissociation kinetics. It can cross the blood-brain barrier (BBB) and induce epichaperome disassembly by non-covalently binding to HSP90, restoring the normal protein-protein interaction network. It is used in research for neurodegenerative diseases like Alzheimer's and cancers such as glioblastoma and metastatic breast cancer.
  • Selective, orally active inhibitor of epichaperomes
  • Crosses the blood-brain barrier (BBB)
  • Induces epichaperome disassembly
  • Restores normal protein-protein interaction networks
  • Reduces neurotoxic protein aggregation
  • Reduces tumor cell survival signals
Catalog No. 50-003-50673 Supplier Medchemexpress LLC Supplier No. 25MGHY137441A
Icapamespib hydrochloride is a selective, orally active inhibitor of Epichaperomes assembled by HSP90 with slow dissociation kinetics. It can cross the blood-brain barrier (BBB) and induce epichaperome disassembly by non-covalently binding to HSP90, restoring the normal protein-protein interaction network. It is used in research for neurodegenerative diseases like Alzheimer's and cancers such as glioblastoma and metastatic breast cancer.
  • Selective, orally active inhibitor of epichaperomes
  • Crosses the blood-brain barrier (BBB)
  • Induces epichaperome disassembly
  • Restores normal protein-protein interaction networks
  • Reduces neurotoxic protein aggregation
  • Reduces tumor cell survival signals