Learn More
Glass Expansion Base for standard quartz torch body for iCAP Q, Glass Expansion
SDP
Catalog No. 501694610
Encompass_Preferred
Base for standard quartz torch body (1231000). Compatible with Thermo Q/RQ/TQ Series ICP-MS.Optional injectors include organics 1.0mm ID (31-808-3440), aqueous 2.0mm ID (31-808-3898), and high TDS 2.5mm ID (31-808-3428). The quartz torch body (50-169-4609) is ideal for most routine organic and aqueous applications but is not for HF matrices. Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-169-4610 Supplier Glass Expansion Supplier No. 318083494
Base for standard quartz torch body (1231000). Compatible with Thermo Q/RQ/TQ Series ICP-MS.Optional injectors include organics 1.0mm ID (31-808-3440), aqueous 2.0mm ID (31-808-3898), and high TDS 2.5mm ID (31-808-3428). The quartz torch body (50-169-4609) is ideal for most routine organic and aqueous applications but is not for HF matrices. Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001