Learn More
Medchemexpress LLC HY-N0014 100mg Medchemexpress, Icariin CAS:489-32-7 Purity:>98%
SDP
Catalog No. 501933354
Encompass_Preferred
Medchemexpress, HY-N0014 100mg Icariin CAS:489-32-7 Icariin is a flavonol glycoside. Icariin inhibits PDE5 and PDE4 activities with IC 50 s of 432 nM and 73.50 μM, respectively. Icariin also is a PPARα activator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3354 Supplier Medchemexpress LLC Supplier No. HYN0014100MG
Medchemexpress, HY-N0014 100mg Icariin CAS:489-32-7 Icariin is a flavonol glycoside. Icariin inhibits PDE5 and PDE4 activities with IC 50 s of 432 nM and 73.50 μM, respectively. Icariin also is a PPARα activator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.