Learn More
Apexbio Technology LLC Icariin 489-32-7 100mg
SDP
Catalog No. 502240311
Encompass_Preferred
Icariin (CAS 489-32-7) is a small-molecule inhibitor targeting phosphodiesterases notably PDE5 and PDE4 It is designed to modulate cyclic nucleotide signaling by inhibiting PDE activity thereby regulating processes such as smooth muscle relaxation and neuronal signaling Icariin exerts its biological activity primarily through the inhibition of PDE5 (IC 0 43 M) and PDE4 (IC 73 50 M) Based on these pharmacological properties icariin holds research potential in the investigation of erectile function nitric oxide synthase expression angiogenesis pathways and osteogenic differentiation in bone marrow stromal cells
This item is not returnable. View return policy
Catalog No. 50-224-0311 Supplier Apexbio Technology LLC Supplier No. N1705100
Icariin (CAS 489-32-7) is a small-molecule inhibitor targeting phosphodiesterases notably PDE5 and PDE4 It is designed to modulate cyclic nucleotide signaling by inhibiting PDE activity thereby regulating processes such as smooth muscle relaxation and neuronal signaling Icariin exerts its biological activity primarily through the inhibition of PDE5 (IC 0 43 M) and PDE4 (IC 73 50 M) Based on these pharmacological properties icariin holds research potential in the investigation of erectile function nitric oxide synthase expression angiogenesis pathways and osteogenic differentiation in bone marrow stromal cells