Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Icariin, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014302 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4302 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4302 Supplier MilliporeSigma Supelco Supplier No. 5660125MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Icariin is a major flavonoid isolated from Epimedium spp. leaves, used as a traditional Chinese medicine to nourish the kidney and reinforce yang.

TRUSTED_SUSTAINABILITY

Specifications

CAS 489-32-7
Physical Form Neat
Linear Formula C33H40O15
MDL Number MFCD00210516
Synonym 4′-O-methyl-8-gamma,gamma-dimethylallylkaempferol-3-rhamnoside-7-glucoside
Type Analytical Standard
Quantity 25 mg
Formula Weight 676.66
Percent Purity ≥95% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.