Learn More
Selleck Chemical LLC Icariin 100mg 489-32-7
SDP
Catalog No. 508245
Encompass_Preferred
Icariin is a cGMP-specific PDE5 inhibitor with IC50 of 0.432 ?M, 167-fold more selective for PDE5 than PDE4. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-824-5 Supplier Selleck Chemical LLC Supplier No. S2312100MG
Icariin is a cGMP-specific PDE5 inhibitor with IC50 of 0.432 ?M, 167-fold more selective for PDE5 than PDE4. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.