Learn More
Medchemexpress LLC Icariside D2 | 38954-02-8 | MFCD29059635 | 1 MG
SDP
Catalog No. 5000420017
Encompass_Preferred
Icariside D2 is a chemical compound isolated from the Annona glabra fruit. It is known for its ability to inhibit angiotensin-converting enzyme and exhibits a range of biological activities.
  • Inhibits angiotensin-converting enzyme
  • Shows cytotoxic activity on HL-60 cells with an IC50 value of 9.0 ± 1.0 μM
  • Induces apoptosis
  • Exhibits anti-inflammatory activity
  • Exhibits anti-HIV activity
Catalog No. 50-004-20017 Supplier Medchemexpress LLC Supplier No. HYN74501MG
Icariside D2 is a chemical compound isolated from the Annona glabra fruit. It is known for its ability to inhibit angiotensin-converting enzyme and exhibits a range of biological activities.
  • Inhibits angiotensin-converting enzyme
  • Shows cytotoxic activity on HL-60 cells with an IC50 value of 9.0 ± 1.0 μM
  • Induces apoptosis
  • Exhibits anti-inflammatory activity
  • Exhibits anti-HIV activity