Learn More
Cayman Chemical IcarIsIde I 10mg
SDP
Catalog No. 502948007
Encompass_Preferred
A flavonoid glycoside; stimulates the proliferation and differentiation of isolated rat osteoblasts in a concentration-dependent manner; increases the cytotoxicity of adriamycin in multidrug-resistant MCF-7/adr breast cancer cells (IC50 = 60.78 µM)
Catalog No. 50-294-8007 Supplier Cayman Chemical Supplier No. 3320910 MG
A flavonoid glycoside; stimulates the proliferation and differentiation of isolated rat osteoblasts in a concentration-dependent manner; increases the cytotoxicity of adriamycin in multidrug-resistant MCF-7/adr breast cancer cells (IC50 = 60.78 µM)