Learn More
TARGETMOL CHEMICALS INC Icariside I 20MG
SDP
Catalog No. 502880087
Encompass_Preferred
Also available in 1 mL and bulk. Please contact Fisher for quotes. Icariside I (Lcariside I) is isolated from Epimedium herb. It can stimulate osteogenic differentiation of BMSCs and inhibit bone resorption activity of osteoclasts. Purity 97.14%
Catalog No. 50-288-0087 Supplier TARGETMOL CHEMICALS INC Supplier No. T380820MG
Add to Cart
Add to Cart
Also available in 1 mL and bulk. Please contact Fisher for quotes. Icariside I (Lcariside I) is isolated from Epimedium herb. It can stimulate osteogenic differentiation of BMSCs and inhibit bone resorption activity of osteoclasts. Purity 97.14%