Learn More
Medchemexpress LLC HY-N1939 5mg Medchemexpress, Icariside I CAS:56725-99-6 Purity:>98%
SDP
Catalog No. 501933862
Encompass_Preferred
Medchemexpress, HY-N1939 5mg Icariside I CAS:56725-99-6 Icariside I is a metabolite of Icarlin, which could regulate bone remodeling and is recognized as an effective agent for the treatment of osteoporosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3862 Supplier Medchemexpress LLC Supplier No. HYN19395MG
Medchemexpress, HY-N1939 5mg Icariside I CAS:56725-99-6 Icariside I is a metabolite of Icarlin, which could regulate bone remodeling and is recognized as an effective agent for the treatment of osteoporosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.