Learn More
Medchemexpress LLC Icatibant acetate | 138614-30-9 | 99.87% | 1364.57 | 1 MG
SDP
Catalog No. 5000346037
Encompass_Preferred
Icatibant acetate is a potent and specific peptide antagonist of the bradykinin B2 receptor. It exhibits an IC50 and Ki of 1.07 nM and 0.798 nM respectively. Icatibant can also potentiate angiotensin III and Lys-des-Arg9-bradykinin.
  • Potent and specific peptide antagonist
  • Targets bradykinin B2 receptor
  • Can potentiate angiotensin III
  • Can potentiate Lys-des-Arg9-bradykinin
Catalog No. 50-003-46037 Supplier Medchemexpress LLC Supplier No. HY10889610MG
Icatibant acetate is a potent and specific peptide antagonist of the bradykinin B2 receptor. It exhibits an IC50 and Ki of 1.07 nM and 0.798 nM respectively. Icatibant can also potentiate angiotensin III and Lys-des-Arg9-bradykinin.
  • Potent and specific peptide antagonist
  • Targets bradykinin B2 receptor
  • Can potentiate angiotensin III
  • Can potentiate Lys-des-Arg9-bradykinin