Learn More
Medchemexpress LLC Icatibant (acetate) | 138614-30-9 | 99.9% | 1364.57 | 10 MG
SDP
Catalog No. 5000346260
Encompass_Preferred
Icatibant acetate (HOE-140 acetate) is a potent and specific peptide antagonist of the bradykinin B2 receptor with an IC50 and Ki of 1.07 nM and 0.798 nM respectively.
  • Potent and specific peptide antagonist of bradykinin B2 receptor.
  • Potentiates angiotensin III and Lys-des-Arg9-bradykinin.
  • Has shown a significant preventive effect on shortening of the large intestine in a mouse model at dosages of 0.3 mg/kg and 1.5 mg/kg.
Catalog No. 50-003-46260 Supplier Medchemexpress LLC Supplier No. HY108896100MG
Icatibant acetate (HOE-140 acetate) is a potent and specific peptide antagonist of the bradykinin B2 receptor with an IC50 and Ki of 1.07 nM and 0.798 nM respectively.
  • Potent and specific peptide antagonist of bradykinin B2 receptor.
  • Potentiates angiotensin III and Lys-des-Arg9-bradykinin.
  • Has shown a significant preventive effect on shortening of the large intestine in a mouse model at dosages of 0.3 mg/kg and 1.5 mg/kg.