Learn More
Medchemexpress LLC Icatibant (acetate) | 138614-30-9 | 99.9% | 1364.57 | 5 ML
SDP
Catalog No. 5000344596
Encompass_Preferred
Icatibant acetate (HOE-140 acetate) is a potent and specific peptide antagonist of the bradykinin B2 receptor, with an IC50 of 1.07 nM and a Ki of 0.798 nM. It is intended for research use only. In in vivo studies, Icatibant has been shown to potentiate angiotensin III and Lys-des-Arg9-bradykinin. Animal studies also demonstrated a significant preventive effect on shortening of the large intestine.
  • Potent and specific peptide antagonist of bradykinin B2 receptor
  • IC50 of 1.07 nM and Ki of 0.798 nM
  • Potentiates angiotensin III and Lys-des-Arg9-bradykinin in vivo
  • Significant preventive effect on shortening of the large intestine in animal models
  • White to off-white solid appearance
  • Soluble in H2O and DMSO at 100 mg/mL
  • Intended for research use only
Catalog No. 50-003-44596 Supplier Medchemexpress LLC Supplier No. HY10889650MG
Icatibant acetate (HOE-140 acetate) is a potent and specific peptide antagonist of the bradykinin B2 receptor, with an IC50 of 1.07 nM and a Ki of 0.798 nM. It is intended for research use only. In in vivo studies, Icatibant has been shown to potentiate angiotensin III and Lys-des-Arg9-bradykinin. Animal studies also demonstrated a significant preventive effect on shortening of the large intestine.
  • Potent and specific peptide antagonist of bradykinin B2 receptor
  • IC50 of 1.07 nM and Ki of 0.798 nM
  • Potentiates angiotensin III and Lys-des-Arg9-bradykinin in vivo
  • Significant preventive effect on shortening of the large intestine in animal models
  • White to off-white solid appearance
  • Soluble in H2O and DMSO at 100 mg/mL
  • Intended for research use only