Learn More
Selleck Chemical LLC Icatibant Acetate
SDP
Catalog No. 5000003269
Encompass_Preferred
Icatibant Acetate(HOE-140 acetate) acts as an antagonist of the bradykinin B2 receptor and is used in the treatment of hereditary angioedema
Catalog No. 50-000-03269 Supplier Selleck Chemical LLC Supplier No. S5695100MG
Icatibant Acetate(HOE-140 acetate) acts as an antagonist of the bradykinin B2 receptor and is used in the treatment of hereditary angioedema