Learn More
Selleck Chemical LLC Icatibant Acetate
SDP
Supplier:  Selleck Chemical LLC S56951G
Encompass_Preferred
Icatibant Acetate(HOE-140 acetate) acts as an antagonist of the bradykinin B2 receptor and is used in the treatment of hereditary angioedema
Catalog No. 50-000-03704