Learn More
Mitegen ICB20 Cryogenic Freezer By IC Biomedical for Sample Storage In Liquid Nitrogen, 20 Liter Capacity
SDP
Catalog No. 5000495036
Encompass
The IC Biomedical ICB20 Cryogenic Freezer is a 21-liter liquid nitrogen freezer designed for use in laboratory, biorepository, and other research applications. Designed to hold biological samples. This unit offers a static holding time of 135 days and has a capacity for up to six 11-inch canisters. The freezer features a rugged, ribbed, high-strength aluminum body with a magniformed necktube design and a durable paint system. Superior vacuum performance and insulation technology ensure maximum holding times. The unit also includes a lockable lid with an enlarged opening for seals or securing ties and universal canister compatibility. The ICB20 has a 2.2-inch (55.9 mm) neck opening, an overall diameter of 14.5 inches (368.3 mm), and a height of 26 inches (660.4 mm). It weighs 26 lbs (11.8 kg) empty and 63 lbs (28.6 kg) when full.
Catalog No. 50-004-95036 Supplier Mitegen Supplier No. TWICB20
The IC Biomedical ICB20 Cryogenic Freezer is a 21-liter liquid nitrogen freezer designed for use in laboratory, biorepository, and other research applications. Designed to hold biological samples. This unit offers a static holding time of 135 days and has a capacity for up to six 11-inch canisters. The freezer features a rugged, ribbed, high-strength aluminum body with a magniformed necktube design and a durable paint system. Superior vacuum performance and insulation technology ensure maximum holding times. The unit also includes a lockable lid with an enlarged opening for seals or securing ties and universal canister compatibility. The ICB20 has a 2.2-inch (55.9 mm) neck opening, an overall diameter of 14.5 inches (368.3 mm), and a height of 26 inches (660.4 mm). It weighs 26 lbs (11.8 kg) empty and 63 lbs (28.6 kg) when full.