Learn More
Mitegen ICB24 Cryogenic Freezer By IC Biomedical for Sample Storages In Liquid Nitrogen, 24 Liter Capacity
SDP
Catalog No. 5000495028
Encompass
The IC Biomedical ICB24 Cryogenic Freezer is designed for laboratory, biorepository, and other research applications requiring liquid nitrogen storage. Designed to hold biological samples. This dewar has a 24-liter liquid nitrogen capacity and an extended static holding time of 218 days. It is built with a rugged, ribbed, high-strength aluminum body, a magniformed necktube design, and a durable paint system. Superior vacuum performance and insulation technology ensure maximum holding times. The unit also features a lockable lid with an enlarged opening for seals or securing ties, and universal canister compatibility, accommodating up to six 11-inch canisters. The ICB24 stands 27.3 inches (693.4 mm) tall, has a neck opening of 2.38 inches (60.4 mm), and an overall diameter of 14.5 inches (368.3 mm). It weighs 26.2 lbs (11.9 kg) empty and 69 lbs (31.3 kg) when full.
Catalog No. 50-004-95028 Supplier Mitegen Supplier No. TWICB24
The IC Biomedical ICB24 Cryogenic Freezer is designed for laboratory, biorepository, and other research applications requiring liquid nitrogen storage. Designed to hold biological samples. This dewar has a 24-liter liquid nitrogen capacity and an extended static holding time of 218 days. It is built with a rugged, ribbed, high-strength aluminum body, a magniformed necktube design, and a durable paint system. Superior vacuum performance and insulation technology ensure maximum holding times. The unit also features a lockable lid with an enlarged opening for seals or securing ties, and universal canister compatibility, accommodating up to six 11-inch canisters. The ICB24 stands 27.3 inches (693.4 mm) tall, has a neck opening of 2.38 inches (60.4 mm), and an overall diameter of 14.5 inches (368.3 mm). It weighs 26.2 lbs (11.9 kg) empty and 69 lbs (31.3 kg) when full.