Learn More
Mitegen ICB47-10 Cryogenic Freezer By IC Biomedical for Sample Storage in Liquid Nitrogen, 48 Liter Capacity
SDP
Catalog No. 5000494985
Encompass
The IC Biomedical ICB47-10 Cryogenic Freezer is designed for laboratory, biorepository, and other research applications. Designed to hold biological samples. This dewar holds up to 48 liters of liquid nitrogen with a static holding time of 123 days. It can fit up to ten 11-inch canisters, offering a large storage capacity. The freezer features a rugged, ribbed, high-strength aluminum body, a magniformed necktube design, and a durable paint system. Superior vacuum performance and insulation technology ensure maximum holding times. The unit also includes a lockable lid with an enlarged opening for seals or securing ties and universal canister compatibility. The ICB47-10 is 26.5 inches (673.1 mm) high, has a 5-inch (127 mm) neck opening, and an overall diameter of 20 inches (508 mm). It weighs 31.5 lbs (14.3 kg) empty and 120 lbs (54 kg) when full.
Catalog No. 50-004-94985 Supplier Mitegen Supplier No. TWICB4710
The IC Biomedical ICB47-10 Cryogenic Freezer is designed for laboratory, biorepository, and other research applications. Designed to hold biological samples. This dewar holds up to 48 liters of liquid nitrogen with a static holding time of 123 days. It can fit up to ten 11-inch canisters, offering a large storage capacity. The freezer features a rugged, ribbed, high-strength aluminum body, a magniformed necktube design, and a durable paint system. Superior vacuum performance and insulation technology ensure maximum holding times. The unit also includes a lockable lid with an enlarged opening for seals or securing ties and universal canister compatibility. The ICB47-10 is 26.5 inches (673.1 mm) high, has a 5-inch (127 mm) neck opening, and an overall diameter of 20 inches (508 mm). It weighs 31.5 lbs (14.3 kg) empty and 120 lbs (54 kg) when full.