Learn More
Mitegen ICB47-6 Cryogenic Freezer By IC Biomedical for Sample Storage in Liquid Nitrogen, 48 Liter Capacity
SDP
Catalog No. 5000495024
Encompass
The IC Biomedical ICB47-6 Cryogenic Freezer is designed for laboratory, biorepository, and other research applications. Designed to hold biological samples. This unit can hold up to 48 liters of liquid nitrogen and features a static hold time of 123 days, with a low evaporation rate of 0.39 liters per day. It comes equipped with six 11-inch canisters and is designed with a rugged, high-strength aluminum body, a magneformed necktube design, and a more durable paint system. Its superior vacuum performance and insulation technology ensure maximum holding times. The lockable lid and universal canister compatibility make it a reliable choice for secure and long-term cryogenic storage.
Catalog No. 50-004-95024 Supplier Mitegen Supplier No. TWICB476
The IC Biomedical ICB47-6 Cryogenic Freezer is designed for laboratory, biorepository, and other research applications. Designed to hold biological samples. This unit can hold up to 48 liters of liquid nitrogen and features a static hold time of 123 days, with a low evaporation rate of 0.39 liters per day. It comes equipped with six 11-inch canisters and is designed with a rugged, high-strength aluminum body, a magneformed necktube design, and a more durable paint system. Its superior vacuum performance and insulation technology ensure maximum holding times. The lockable lid and universal canister compatibility make it a reliable choice for secure and long-term cryogenic storage.