Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICBA 98.5+%, TCI America™
SDP

Catalog No. I090050MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I090050MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I090050MG Supplier TCI America Supplier No. I090050MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1207461-57-1
Molecular Formula C78H16
Molecular Weight (g/mol) 952.986
MDL Number MFCD20264904
InChI Key HQCAMIYWHBVPQE-UHFFFAOYSA-N
PubChem CID 91972098
SMILES C1C2C3=CC=CC=C3C1C45C26C7=C8C9=C1C2=C3C8=C8C6=C6C4=C4C%10=C%11C%12=C%13C%14=C%15C%16=C%17C(=C1C1=C2C2=C%18C%19=C%20C2=C3C8=C2C%20=C(C%10=C26)C2=C%11C3=C%13C%15=C6C%17=C1C%181C6(C3=C2%19)C2CC1C1=CC=CC=C21)C1=C%16C2=C%14C(=C%124)C5=C2C7=C19

Specifications

Melting Point 260°C
Color Black
Quantity 50 mg
Formula Weight 952.99
Percent Purity ≥98.5% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material ICBA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.