Learn More
Cayman Chemical ICBP112 PosItIve CNTRL 24nmol
SDP
Catalog No. 502893689
Encompass_Preferred
An immunosuppressant efficacious and preventative in a rat model of complete Freund’s adjuvant-induced polyarthritis at 200 and 100 mg/kg respectively 50 and 90 protective against EAE in rats at 100 and 400 mg/kg respectively
Catalog No. 50-289-3689 Supplier Cayman Chemical Supplier No. 60085324 NMOL
An immunosuppressant efficacious and preventative in a rat model of complete Freund’s adjuvant-induced polyarthritis at 200 and 100 mg/kg respectively 50 and 90 protective against EAE in rats at 100 and 400 mg/kg respectively