Learn More
Medchemexpress LLC ICCB-19 (hydrochloride) | 1803605-68-6 | 99.3% | 291.84 | C12H22ClN3OS | 5 MG
SDP
Catalog No. 5000221100
Encompass_Preferred
ICCB-19 hydrochloride is a research-grade small molecule that inhibits TRADD-mediated signaling and indirectly inhibits RIPK1 kinase activity. It has been reported to induce autophagy, modulate TNF-induced inflammatory responses, and affect RIPK1-dependent apoptosis in cellular and animal models.
  • Targets TRADD and indirectly inhibits RIPK1 kinase activity.
  • Binds TRADD N-terminal domain to disrupt protein interactions.
  • Promotes autophagy via K63-linked ubiquitination of Beclin-1.
  • Reduces TNF-induced inflammatory gene expression in cell and animal studies.
  • Provided as a hydrochloride salt with reported high purity.
  • Available as solid and DMSO solution aliquots for research use.
Catalog No. 50-002-21100 Supplier Medchemexpress LLC Supplier No. HY1387795MG
ICCB-19 hydrochloride is a research-grade small molecule that inhibits TRADD-mediated signaling and indirectly inhibits RIPK1 kinase activity. It has been reported to induce autophagy, modulate TNF-induced inflammatory responses, and affect RIPK1-dependent apoptosis in cellular and animal models.
  • Targets TRADD and indirectly inhibits RIPK1 kinase activity.
  • Binds TRADD N-terminal domain to disrupt protein interactions.
  • Promotes autophagy via K63-linked ubiquitination of Beclin-1.
  • Reduces TNF-induced inflammatory gene expression in cell and animal studies.
  • Provided as a hydrochloride salt with reported high purity.
  • Available as solid and DMSO solution aliquots for research use.