Learn More
Selleck Chemical LLC ICCB-19 hydrochloride
SDP
Supplier:  Selleck Chemical LLC S607810MM/1ML
Encompass_Preferred
ICCB-19 hydrochloride is an inhibitor of TNFRSF1A Associated Via Death Domain (TRADD) with IC50 of 1 12 M and 2 01 M for protecting Velcade-induced apoptosis in Jurkat cells and protecting RDA in MEFs respectively ICCB-19 indirectly inhibits Receptor-interacting serine/threonine-protein kinase 1 (RIPK1) ICCB-19 effectively induces autophagy
Catalog No. 50-000-00782