Learn More
Enzo Life Sciences SCREEN-WELL ICCB Known Bioactives library (100µl/well)
SDP
Catalog No. 502010217
Encompass_Preferred
The ICCB Known Bioactives Library is an important product for use in chemical genetics and drug discovery. The library is a collection of 472 diverse biologically active compounds with defined biological activity and was developed in collaboration with the Harvard Institute of Chemistry and Cell Biology. It includes the following classes of compounds: GPCR ligands, Second messenger modulators, Nuclear receptor ligands, Actin & tubulin modulators, Kinase inhibitors, Protease inhibitors, Ion channel blockers, Gene regulation agents, Lipid biosynthesis inhibitors, many other classes. Each compound is supplied dissolved in DMSO at 100µL and is ready for assaying. The library can be used for assay development and validation, mechanism profiling, lead screening or as a reference library. Long Term Storage: -80°C. Applications: HTS .
Catalog No. 50-201-0217 Supplier Enzo Life Sciences Supplier No. BML28400100
Add to Cart
Add to Cart
Request a Quote
The ICCB Known Bioactives Library is an important product for use in chemical genetics and drug discovery. The library is a collection of 472 diverse biologically active compounds with defined biological activity and was developed in collaboration with the Harvard Institute of Chemistry and Cell Biology. It includes the following classes of compounds: GPCR ligands, Second messenger modulators, Nuclear receptor ligands, Actin & tubulin modulators, Kinase inhibitors, Protease inhibitors, Ion channel blockers, Gene regulation agents, Lipid biosynthesis inhibitors, many other classes. Each compound is supplied dissolved in DMSO at 100µL and is ready for assaying. The library can be used for assay development and validation, mechanism profiling, lead screening or as a reference library. Long Term Storage: -80°C. Applications: HTS .