Learn More
Medchemexpress LLC HY-134333 5mg Medchemexpress, ICCB280 CAS:2041072-41-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1343335MG
Encompass_Preferred
Medchemexpress, HY-134333 5mg ICCB280 CAS:2041072-41-5 ICCB280 is a potent inducer of C/EBPα. ICCB280 exhibits anti-leukemic properties including terminal differentiation, proliferation arrest, and apoptosis through activation of C/EBPα and affecting its downstream targets (such as C/EBPε, G-CSFR and c-Myc). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1363