Learn More
Tcp Reliable Inc Flexible Ice Blanket 12mL Cell 12W x 8L: 16.85in x 23.30in.
Catalog No. NC0651696
Encompass_Preferred
This item has a minimum order quantity of 3 cases per supplier requirements. Flexible Ice Blanket 12mL Cell 12W x 8L: 16.85in x 23.30in. The Flexible Ice Blanket™ offers a design that can wrap around a payload, even when frozen, providing enhanced thermal protection of temperature-sensitive products. Ideal for a variety of applications, these blankets have a freezing/melting point of 0°C and stay cold without leaking or spilling.
Catalog No. NC0651696 Supplier Tcp Reliable Inc Supplier No. 85043
This item has a minimum order quantity of 3 cases per supplier requirements. Flexible Ice Blanket 12mL Cell 12W x 8L: 16.85in x 23.30in. The Flexible Ice Blanket™ offers a design that can wrap around a payload, even when frozen, providing enhanced thermal protection of temperature-sensitive products. Ideal for a variety of applications, these blankets have a freezing/melting point of 0°C and stay cold without leaking or spilling.