Learn More
Polar Tech Industries Ice Brix 6oz 5 x 4 x ", IB 6 (96/CS)
SDP
Catalog No. NC1557979
Encompass_Preferred
This item has a minimum qty of 3 per supplier requirements.
Polar Tech's leakproof Ice Brix are the industry's original, and truly, leakproof gel pack. Each Ice Brix is made with nontoxic biodegradable gel and is listed as FDA GRAS. The gel is sealed in a heavy duty poly bag and provides exceptional temperature control and protection. Don't worry about gel destroying labels, boxes, or the perception of toxic gels leeching into your products again The Ice Brix brand is the best.
Catalog No. NC1557979 Supplier Polar Tech Industries Supplier No. IB 6
This item has a minimum qty of 3 per supplier requirements.
Polar Tech's leakproof Ice Brix are the industry's original, and truly, leakproof gel pack. Each Ice Brix is made with nontoxic biodegradable gel and is listed as FDA GRAS. The gel is sealed in a heavy duty poly bag and provides exceptional temperature control and protection. Don't worry about gel destroying labels, boxes, or the perception of toxic gels leeching into your products again The Ice Brix brand is the best.