Learn More
Chemglass Life Sciences Chemglass 2.5 L Ice Bucket with Lid, Black
SDP
Catalog No. NC1837040
Encompass_Preferred
A durable 2.5 L laboratory ice bucket with a snug-fitting lid, designed to keep samples and reagents cold while minimizing condensation. Constructed from expanded urethane for wide temperature performance and easy handling.
  • 2.5 L capacity.
  • Expanded urethane construction for temperature resilience.
  • Temperature range -196°C to 100°C.
  • Snug-fitting lid reduces evaporation.
  • Lightweight, durable, and non-sweating.
  • Pour spout, drip-proof rims, and fill line indicator.
  • Lightly textured non-slip finish for secure transport.
Catalog No. NC1837040 Supplier Chemglass Life Sciences Supplier No. CLS17099202
A durable 2.5 L laboratory ice bucket with a snug-fitting lid, designed to keep samples and reagents cold while minimizing condensation. Constructed from expanded urethane for wide temperature performance and easy handling.
  • 2.5 L capacity.
  • Expanded urethane construction for temperature resilience.
  • Temperature range -196°C to 100°C.
  • Snug-fitting lid reduces evaporation.
  • Lightweight, durable, and non-sweating.
  • Pour spout, drip-proof rims, and fill line indicator.
  • Lightly textured non-slip finish for secure transport.