Learn More
Chemglass Life Sciences Chemglass Ice Bucket with Lid, 4 L, Blue
SDP
Catalog No. NC2959221
Encompass_Preferred
Expanded urethane laboratory ice bucket for cold storage and handling of wet ice, dry ice, salt slurries, and other ultra-low temperature materials. Includes a snug-fitting lid that stores under the base, a pour spout, drip-proof rims, a fill-line indicator, and a lightly textured non-slip finish for secure transport.
  • Superior insulating properties and high chemical resistance.
  • Compatible with wet ice, dry ice, salt slurries, and liquid nitrogen.
  • Impervious to moisture and odors.
  • Includes a snug-fitting lid that stores under base.
  • Features pour spout, drip-proof rims, and fill-line indicator.
  • Lightly textured non-slip finish and raised edge for secure handling.
  • 4.0 L capacity; diameter 10.56 in and height with lid 8.93 in.
Catalog No. NC2959221 Supplier Chemglass Life Sciences Supplier No. CLS17099401
Expanded urethane laboratory ice bucket for cold storage and handling of wet ice, dry ice, salt slurries, and other ultra-low temperature materials. Includes a snug-fitting lid that stores under the base, a pour spout, drip-proof rims, a fill-line indicator, and a lightly textured non-slip finish for secure transport.
  • Superior insulating properties and high chemical resistance.
  • Compatible with wet ice, dry ice, salt slurries, and liquid nitrogen.
  • Impervious to moisture and odors.
  • Includes a snug-fitting lid that stores under base.
  • Features pour spout, drip-proof rims, and fill-line indicator.
  • Lightly textured non-slip finish and raised edge for secure handling.
  • 4.0 L capacity; diameter 10.56 in and height with lid 8.93 in.