Learn More
Cole-Parmer Cryopak Ice-Pak Cold Packs, 10-1/2" x 8" x 1-1/2", 12/cs
Supplier:  Cole-Parmer 0634540
Encompass_Preferred
Rigid packs freeze solid in 6 to 10 hours and remain at constant 20°>F (-6°>C) temperature for up to 72 hours. Packs are nontoxic and won't contaminate samples. Use with insulated shippers and containers for best results when shipping heat-sensitive materials.
Catalog No. NC2197690