Learn More
Epredia™ Ice-It™ Freezing Spray
Catalog No. 22050319 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Ice-It Freezing Spray
Catalog No. Product Type
22-050-319 Ice-It Freezing Spray
1 options
This item is not returnable. View return policy
Catalog No. 22-050-319 Supplier Epredia™ Supplier No. 8325
Add to Cart
Add to Cart
This item is not returnable. View return policy
Instantly cool or freeze samples at the microtome or cryostat with the ozone-friendly Ice-It™ Freezing Spray.
  • Variable pressure trigger and capillary nozzle allow for accurate cooling or freezing of tissue
  • No CFC's or HFC's, so it will not deplete stratospheric ozone
Specifications
Description 11 oz. can; 12/Cs
Product Type Ice-It Freezing Spray
WARNING: Reproductive Harm - www.P65Warnings.ca.gov