Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Ice Melting Kit
GREENER_CHOICESCI_ED

Catalog No. S139434 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
6+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139434 6+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139434 Supplier Supertek™ Supplier No. PH20095
Add to Cart
Add to Cart
This item is not returnable. View return policy

Demonstrate the difference in thermal conductivity of two different materials with this Supertek™ Ice Melting Kit. The kit consists of two identical blocks with one made of aluminum and other of plastic.

  • Demonstration: When an ice cube is placed on aluminum it turns into water in seconds while on plastic block it melts slowly.
  • Why: The aluminum block has a larger heat capacity and a higher thermal conductivity while the plastic block has lower values of both parameters.
  • The kit consists of two identical blocks one made of aluminum and the other of plastic.
  • Comes with instructions.

Specifications

Grade 6+
Format Kit
Languages English
Subject Physical Science - Heat
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.