Learn More
Cosmo Bio Usa Inc Iceon Type 1 Cooling Rack, Holds 24 x 1.5ml tubes
SDP
Catalog No. NC0282616
Encompass
Faster Refrigeration - Tubes will not be touched directly with water and ice - Made of aluminum and its surface is processed not to be rusted for the long time use. - Compactly designed just like putting into the small bucket. - Can be piled on and in a refrigerator or freezer, they can be used as racks of sample tubes. Type 1 Feature:- Can contain 24 tubes of 1.5 ml - Numbers (1-8) and alphabets (A to C) are inscribed on the surface. Easy to be sortedout in order.
Catalog No. NC0282616 Supplier Cosmo Bio Usa Inc Supplier No. SKBIO1
Faster Refrigeration - Tubes will not be touched directly with water and ice - Made of aluminum and its surface is processed not to be rusted for the long time use. - Compactly designed just like putting into the small bucket. - Can be piled on and in a refrigerator or freezer, they can be used as racks of sample tubes. Type 1 Feature:- Can contain 24 tubes of 1.5 ml - Numbers (1-8) and alphabets (A to C) are inscribed on the surface. Easy to be sortedout in order.