Learn More
Hach Company Ice-Pic 0.5NTU Cal/Veri Module
Catalog No. 5000207172 Shop All Hach Company Products
Encompass
Ice-Pic 0 5 NTU Calibration / Verification ModuleCan be used with the 1720C and D instruments Verify calibration in less than 60 seconds No loss of sample flow to complete a verification and no sample cell to recondition after verification
Catalog No. 50-002-07172 Supplier Hach Company Supplier No. 5222500
Ice-Pic 0 5 NTU Calibration / Verification ModuleCan be used with the 1720C and D instruments Verify calibration in less than 60 seconds No loss of sample flow to complete a verification and no sample cell to recondition after verification