Learn More
Hach Company Ice-Pic 20NTU Cal/Veri Module
Supplier:  Hach Company 5225000
Encompass
Ice-Pic 20 NTUCalibration / Verification ModuleVerify calibration in less than 60 seconds Available in 1 and 20 NTU modules Can be used with the 1720E D and C instruments No loss of sample flow to complete a verification No sample cell to recondition after verification
Catalog No. 50-002-07131