Learn More
Julabo USA Inc Each, Icing protection/sleeve for pump connectors.
SDP
Catalog No. 507534165
Encompass_Preferred
Bath Accessories, Julabo, Icing protection/sleeve for pump connectors. Install on pump and fitting connections to reduce ice build up / heat loss. Insulation with adjustable Velcro straps.
Catalog No. 50-753-4165 Supplier Julabo USA Inc Supplier No. 8970750
Bath Accessories, Julabo, Icing protection/sleeve for pump connectors. Install on pump and fitting connections to reduce ice build up / heat loss. Insulation with adjustable Velcro straps.