Learn More
Felix Storch Inc Ice & Water Dispenser 160 lb. Daily Ice Production
SDP
Catalog No. NC3901160
Encompass_Preferred
The AIWD160 is ETL-S listed to NSF-12 commercial standards. Measuring just 17.5 inches from the front leg to the unit rear makes this a perfect fit on standard counters, with a slim width under 14 inches. This unit can produce up to 160 pounds of ice per day. Three dispenser functions - ice, water, or ice+water.Adjustable legs offer flexible height options, with a 304 grade stainless steel front complemented by the black dispenser panel and metal cabinet. The removable tray and sealed back make for easy cleaning. This unit comes equipped with UV lighting in the dispenser. The AIWD160 must be used with a drain and should be placed in a space that allows for draining below the unit. It is top vented. AIWD160 is the perfect unit for hospital waiting areas, break rooms, or similar settings looking for a water and ice dispenser that is attractive and efficient.Note: A water filtration system is available and recommended for optimal performance of the unit.
Catalog No. NC3901160 Supplier Felix Storch Inc Supplier No. AIWD160
Add to Cart
Add to Cart
Request a Quote
The AIWD160 is ETL-S listed to NSF-12 commercial standards. Measuring just 17.5 inches from the front leg to the unit rear makes this a perfect fit on standard counters, with a slim width under 14 inches. This unit can produce up to 160 pounds of ice per day. Three dispenser functions - ice, water, or ice+water.Adjustable legs offer flexible height options, with a 304 grade stainless steel front complemented by the black dispenser panel and metal cabinet. The removable tray and sealed back make for easy cleaning. This unit comes equipped with UV lighting in the dispenser. The AIWD160 must be used with a drain and should be placed in a space that allows for draining below the unit. It is top vented. AIWD160 is the perfect unit for hospital waiting areas, break rooms, or similar settings looking for a water and ice dispenser that is attractive and efficient.Note: A water filtration system is available and recommended for optimal performance of the unit.