Learn More
Felix Storch Inc Ice and Water Dispenser 220 lb. Daily Ice Production
SDP
Catalog No. NC3901153
Encompass_Preferred
Accucold's series of commercially approved ice machines and countertop ice makers bring powerful performance, user-friendly ENERGY STAR certified countertop ice + water dispenser that is ETL-S listed to NSF-12 standards. This unit can produce up to 220 lbs. of nugget ice daily, and the interior can store up to 6.2 lbs. It features three easy-to-use functions - ice, water, or ice + water, which provide expanded functionality, while the illuminated display and digital controls ensure easy access and control of the unit. This unit ships with a filter kit.Adjustable legs offer flexible height options, while the stainless steel front provides a professional look. The removable tray and sealed back make for easy cleaning. This unit comes equipped with UV lighting in the dispenser and is top-vented.The AIWD220FLTR is the perfect unit for hospital waiting areas, break rooms, or similar settings looking for a water and ice dispenser that is attractive, efficient, and user-friendly.
Catalog No. NC3901153 Supplier Felix Storch Inc Supplier No. AIWD220FLTR
Add to Cart
Add to Cart
Request a Quote
Accucold's series of commercially approved ice machines and countertop ice makers bring powerful performance, user-friendly ENERGY STAR certified countertop ice + water dispenser that is ETL-S listed to NSF-12 standards. This unit can produce up to 220 lbs. of nugget ice daily, and the interior can store up to 6.2 lbs. It features three easy-to-use functions - ice, water, or ice + water, which provide expanded functionality, while the illuminated display and digital controls ensure easy access and control of the unit. This unit ships with a filter kit.Adjustable legs offer flexible height options, while the stainless steel front provides a professional look. The removable tray and sealed back make for easy cleaning. This unit comes equipped with UV lighting in the dispenser and is top-vented.The AIWD220FLTR is the perfect unit for hospital waiting areas, break rooms, or similar settings looking for a water and ice dispenser that is attractive, efficient, and user-friendly.