Learn More
Felix Storch Inc Ice & Water Dispenser 282 lb. Daily Ice Production
SDP
Catalog No. NC3901167
Encompass_Preferred
The AIWD282 is ETL-S listed to NSF-12 commercial standards. Measuring 18 3/4 inches from the front leg to the unit rear makes this a perfect fit on standard counters. This unit can produce up to 282 pounds of ice per day. Three dispenser functions - ice, water, or ice+waterAdjustable legs offer flexible height options, while the stainless steel front provides a professional look. The removable tray and sealed back make for easy cleaning. This unit comes equipped with UV lighting in the dispenser. The AIWD282 must be used with a drain and should be placed in a space that allows for draining below the unit. It is top vented.With its countertop footprint and large production output, the AIWD282 is the perfect unit for hospital waiting areas, break rooms, or similar settings looking for a water and ice dispenser that is attractive and efficient.Note: A water filtration system is available and recommended for optimal performance of the unit. See FILTER282TWIN.
Catalog No. NC3901167 Supplier Felix Storch Inc Supplier No. AIWD282
Add to Cart
Add to Cart
Request a Quote
The AIWD282 is ETL-S listed to NSF-12 commercial standards. Measuring 18 3/4 inches from the front leg to the unit rear makes this a perfect fit on standard counters. This unit can produce up to 282 pounds of ice per day. Three dispenser functions - ice, water, or ice+waterAdjustable legs offer flexible height options, while the stainless steel front provides a professional look. The removable tray and sealed back make for easy cleaning. This unit comes equipped with UV lighting in the dispenser. The AIWD282 must be used with a drain and should be placed in a space that allows for draining below the unit. It is top vented.With its countertop footprint and large production output, the AIWD282 is the perfect unit for hospital waiting areas, break rooms, or similar settings looking for a water and ice dispenser that is attractive and efficient.Note: A water filtration system is available and recommended for optimal performance of the unit. See FILTER282TWIN.