Learn More
Cellular Dynamics International iCell Cardiomyocytes, human iPSC-derived, 3 vials, ≥ 1.0 x 10^6 viable cells each, >95% cTNT-positive, cryopreserved single-cell suspension
Catalog No. NC2380344 Shop All Cellular Dynamics International Products
Encompass
iCell Cardiomyocytes, human iPSC-derived, 3 vials, ≥ 1.0 x 10^6 viable cells each, >95% cTNT-positive, cryopreserved single-cell suspension
Catalog No. NC2380344 Supplier Cellular Dynamics International Supplier No. R1117
iCell Cardiomyocytes, human iPSC-derived, 3 vials, ≥ 1.0 x 10^6 viable cells each, >95% cTNT-positive, cryopreserved single-cell suspension