Learn More
Cellular Dynamics International iCell Cardiomyocytes DCM LMNA L35P, Cryopreserved single-cell suspensions, ≥ 4 x 10^6 cells/vial, >95% cardiomyocytes
Supplier: Cellular Dynamics International C1016
FUJIFILM IRVINE SCIENTIFIC's iCell Cardiomyocytes are terminally differentiated, cryopreserved human iPSC-derived cardiomyocytes specifically modeled for Dilated Cardiomyopathy (DCM) with an LMNA L35P genetic mutation. These cells reproduce physiological characteristics of native human cardiomyocytes, making them suitable for cardiac research applications such as drug discovery and disease modeling.
- Cell type: Cardiomyocytes
- Cell source: Human iPSCs
- Disease model: Dilated Cardiomyopathy (DCM)
- Genetic mutation: LMNA L35P
- Format: Cryopreserved single-cell suspensions
- Quantity per vial: ≥ 4 x 10^6 cells/vial
- Purity: >95% cardiomyocytes
- Assay readiness: 4 days post-thaw
- Storage: Vapor phase of liquid nitrogen
- Applications: Disease modeling, drug discovery, cardiotoxicity screening, regenerative medicine
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.